siRNA Details
| Gene Symbol: FLJ27354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033981.1 | 1 | 132 | |||
| GenBank mRNA | AK130864.1 | - | 1 | 132 | ||
| Gene Symbol: LRRC8C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017002503.1 | XP_016857992.1 | 1 | 59 | ||

FLJ27354
Genomic Map