siRNA Details
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 1 | 324 | ||
XM_011528562.2 | XP_011526864.1 | 1 | 318 | |||
XM_011528563.1 | XP_011526865.1 | 1 | 318 | |||
GenBank mRNA | BC023657.2 | - | 1 | 324 | ||
BC065483.1 | - | 1 | 324 | |||
BQ067609.1 | - | 1 | 79 |
