siRNA Details
| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006690.3 | NP_006681.1 | 9 | 3707 | ||
| XM_011528500.2 | XP_011526802.1 | 8 | 3437 | |||
| XM_017027597.1 | XP_016883086.1 | 8 | 3542 | |||
| XM_017027598.1 | XP_016883087.1 | 9 | 3678 | |||
| GenBank mRNA | BC047614.1 | - | 9 | 3647 | ||
| Gene Symbol: MMP24-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102705.1 | 2 | 1703 | |||
| NR_102706.1 | 2 | 1666 | ||||
| GenBank mRNA | CN479177.1 | - | 1 | 59 | ||

MMP24
Genomic Map