siRNA Details
| Gene Symbol: BCRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037566.1 | 4 | 488 | |||
| GenBank mRNA | AK124943.1 | - | 4 | 488 | ||
| Gene Symbol: LOC102724751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017030164.1 | XP_016885653.1 | 1 | 21 | ||
| XM_017030165.1 | XP_016885654.1 | 1 | 21 | |||

BCRP2
Genomic Map