siRNA Details
| Gene Symbol: FLJ41327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123321.1 | - | 1 | 2015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ41327| Gene Symbol: FLJ41327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123321.1 | - | 1 | 2015 | ||