siRNA Details
| Gene Symbol: LOC401052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008737.1 | NP_001008737.1 | 4 | 929 | ||
| GenBank mRNA | AK022260.1 | - | 4 | 929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401052| Gene Symbol: LOC401052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008737.1 | NP_001008737.1 | 4 | 929 | ||
| GenBank mRNA | AK022260.1 | - | 4 | 929 | ||