siRNA Details
| Gene Symbol: KRBOX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122033.1 | 1 | 521 | |||
| GenBank mRNA | AK057338.1 | - | 1 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRBOX1-AS1| Gene Symbol: KRBOX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122033.1 | 1 | 521 | |||
| GenBank mRNA | AK057338.1 | - | 1 | 521 | ||