siRNA Details
| Gene Symbol: XCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017006196.1 | XP_016861685.1 | 4 | 5409 | ||
| XM_017006197.1 | XP_016861686.1 | 6 | 5661 | |||
| XM_017006198.1 | XP_016861687.1 | 5 | 5474 | |||
| XM_017006199.1 | XP_016861688.1 | 4 | 4471 | |||
| XM_017006200.1 | XP_016861689.1 | 3 | 5329 | |||
| XM_017006201.1 | XP_016861690.1 | 2 | 5178 | |||
| GenBank mRNA | AK092973.1 | - | 1 | 2885 | ||

XCR1
Genomic Map