siRNA Details
| Gene Symbol: LINC01205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109841.1 | 1, 2 | 218 | |||
| NR_109842.1 | 1, 2 | 218 | ||||
| GenBank mRNA | AK058092.1 | - | 1, 2 | 218 | ||
| BC044821.1 | - | 1, 2 | 49 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01205| Gene Symbol: LINC01205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109841.1 | 1, 2 | 218 | |||
| NR_109842.1 | 1, 2 | 218 | ||||
| GenBank mRNA | AK058092.1 | - | 1, 2 | 218 | ||
| BC044821.1 | - | 1, 2 | 49 | |||