siRNA Details
| Gene Symbol: MBNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314057.1 | NP_001300986.1 | 1 | 76 | ||
| NM_021038.4 | NP_066368.2 | 1 | 76 | |||
| NM_207292.2 | NP_997175.1 | 1 | 76 | |||
| GenBank mRNA | AB007888.1 | - | 1 | 76 | ||
| AI971426.1 | - | 1 | 483 | |||
| BM686432.1 | - | 1 | 453 | |||
| Gene Symbol: MBNL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027037.1 | 2 | 714 | |||
| NR_027038.1 | 2 | 725 | ||||
| GenBank mRNA | AK304857.1 | - | 2 | 634 | ||
| AK316515.1 | - | 2 | 654 | |||

MBNL1
Genomic Map