siRNA Details
Gene Symbol: LINC00885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034088.1 | 2 | 486 | |||
GenBank mRNA | AK129507.1 | - | 2 | 486 | ||
BC151150.1 | - | 2 | 367 | 9021323 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034088.1 | 2 | 486 | |||
GenBank mRNA | AK129507.1 | - | 2 | 486 | ||
BC151150.1 | - | 2 | 367 | 9021323 |