siRNA Details
| Gene Symbol: ZNF732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 1126 | ||
| GenBank mRNA | AK302099.1 | - | 3 | 1126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF732| Gene Symbol: ZNF732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 1126 | ||
| GenBank mRNA | AK302099.1 | - | 3 | 1126 | ||