siRNA Details
| Gene Symbol: PRR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214711.3 | NP_999876.2 | 3 | 814 | ||
| GenBank mRNA | BX640892.1 | - | 3 | 727 | ||
| BX640924.1 | - | 3 | 765 | |||
| BX640997.1 | - | 3 | 801 | |||
| BX647183.1 | - | 4 | 638 | |||
| BX647185.1 | - | 5 | 744 | |||
| BX648724.1 | - | 3 | 763 | |||
| BX648872.1 | - | 3 | 686 | |||
| BX648895.1 | - | 3 | 763 | |||
| BX648896.1 | - | 4 | 698 | |||

PRR27
Genomic Map