siRNA Details
| Gene Symbol: LINC01060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095968.1 | - | 3, 4 | 258 | ||
| AY494056.1 | - | 3, 4 | 356 | |||
| DA579097.1 | - | 3, 4 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01060| Gene Symbol: LINC01060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095968.1 | - | 3, 4 | 258 | ||
| AY494056.1 | - | 3, 4 | 356 | |||
| DA579097.1 | - | 3, 4 | 259 | |||