siRNA Details
| Gene Symbol: MGC39584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038377.1 | 4, 5 | 684 | |||
| GenBank mRNA | BC029568.1 | - | 4, 5 | 684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC39584| Gene Symbol: MGC39584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038377.1 | 4, 5 | 684 | |||
| GenBank mRNA | BC029568.1 | - | 4, 5 | 684 | ||