siRNA Details
| Gene Symbol: C5orf63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164478.1 | NP_001157950.1 | 4 | 287 | ||
| NM_001164479.1 | NP_001157951.1 | 4 | 287 | |||
| XM_017009464.1 | XP_016864953.1 | 3 | 271 | |||
| XM_017009465.1 | XP_016864954.1 | 3 | 271 | |||
| GenBank mRNA | AK126569.1 | - | 5 | 385 | ||
| BQ269742.1 | - | 2 | 422 | |||
| DA507232.1 | - | 4 | 287 | |||

C5orf63
Genomic Map