siRNA Details
Gene Symbol: AACSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024035.2 | 4 | 626 | |||
NR_135095.1 | 4 | 626 | ||||
GenBank mRNA | BC140764.1 | - | 4 | 577 | 9021632 | |
BC144474.1 | - | 4 | 576 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AACSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024035.2 | 4 | 626 | |||
NR_135095.1 | 4 | 626 | ||||
GenBank mRNA | BC140764.1 | - | 4 | 577 | 9021632 | |
BC144474.1 | - | 4 | 576 |