siRNA Details
| Gene Symbol: HTATSF1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033884.1 | 1 | 2797 | |||
| GenBank mRNA | AK128391.1 | - | 3 | 1164 | ||
| BX648511.1 | - | 2 | 1305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTATSF1P2| Gene Symbol: HTATSF1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033884.1 | 1 | 2797 | |||
| GenBank mRNA | AK128391.1 | - | 3 | 1164 | ||
| BX648511.1 | - | 2 | 1305 | |||