siRNA Details
| Gene Symbol: STMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190766.1 | NP_001177695.1 | 5 | 667 | ||
| GenBank mRNA | AK026805.1 | - | 2 | 1430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STMND1| Gene Symbol: STMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190766.1 | NP_001177695.1 | 5 | 667 | ||
| GenBank mRNA | AK026805.1 | - | 2 | 1430 | ||