siRNA Details
| Gene Symbol: TRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012288.3 | NP_036420.1 | 1 | 219 | ||
| GenBank mRNA | AB621814.1 | - | 1 | 215 | ||
| AK291751.1 | - | 1 | 105 | |||
| BC028121.1 | - | 1 | 219 | 5169808 | ||
| D31762.1 | - | 1 | 145 | |||
| KU178488.1 | - | 1 | 70 | |||
| KU178489.1 | - | 1 | 70 | |||

TRAM2
Genomic Map