siRNA Details
| Gene Symbol: TRAM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103446.1 | 1 | 393 | |||
| NR_103447.1 | 1 | 297 | ||||
| GenBank mRNA | AK095117.1 | - | 1 | 288 | ||
| BE741421.1 | - | 1 | 109 | |||
| DB479639.1 | - | 1 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAM2-AS1| Gene Symbol: TRAM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103446.1 | 1 | 393 | |||
| NR_103447.1 | 1 | 297 | ||||
| GenBank mRNA | AK095117.1 | - | 1 | 288 | ||
| BE741421.1 | - | 1 | 109 | |||
| DB479639.1 | - | 1 | 299 | |||