siRNA Details
| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 5 | 1045 | |||
| GenBank mRNA | AK096288.1 | - | 5 | 1044 | ||
| AK291169.1 | - | 6 | 1149 | |||
| Gene Symbol: ZNF890P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK295403.1 | - | 6 | 1039 | ||

ZNF815P
Genomic Map