siRNA Details
| Gene Symbol: LOC401324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038864.1 | 3 | 2230 | |||
| GenBank mRNA | AL832092.1 | - | 3 | 2230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401324| Gene Symbol: LOC401324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038864.1 | 3 | 2230 | |||
| GenBank mRNA | AL832092.1 | - | 3 | 2230 | ||