siRNA Details
| Gene Symbol: MAGI2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038343.2 | 1 | 197 | |||
| NR_038344.1 | 1 | 197 | ||||
| GenBank mRNA | AK311103.1 | - | 1 | 165 | ||
| BI521274.1 | - | 1 | 197 | |||
| CN311982.1 | - | 1 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGI2-AS3| Gene Symbol: MAGI2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038343.2 | 1 | 197 | |||
| NR_038344.1 | 1 | 197 | ||||
| GenBank mRNA | AK311103.1 | - | 1 | 165 | ||
| BI521274.1 | - | 1 | 197 | |||
| CN311982.1 | - | 1 | 164 | |||