siRNA Details
| Gene Symbol: RAB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008749.2 | NP_001008749.2 | 3, 4 | 577 | ||
| XM_005249996.3 | XP_005250053.1 | 3, 4 | 860 | |||
| XM_005249997.3 | XP_005250054.1 | 3, 4 | 843 | |||
| XM_005249998.3 | XP_005250055.1 | 3, 4 | 838 | |||
| XM_017012207.1 | XP_016867696.1 | 3, 4 | 773 | |||
| GenBank mRNA | BC140796.1 | - | 2, 3 | 447 | 9021664 | |
| BC171745.1 | - | 2, 3 | 447 | |||
| DN996121.1 | - | 3, 4 | 551 | |||

RAB19
Genomic Map