siRNA Details
| Gene Symbol: OR2A20P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002158.1 | 1 | 907 | |||
| GenBank mRNA | AK172840.1 | - | 1 | 2261 | ||
| BC016940.1 | - | 3 | 1335 | |||
| Gene Symbol: OR2A9P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002157.1 | 1 | 759 | |||

OR2A20P
Genomic Map