siRNA Details
| Gene Symbol: SNX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012994.1 | NP_001013012.1 | 5 | 885 | ||
| XM_011518691.2 | XP_011516993.1 | 5 | 922 | |||
| XM_017014716.1 | XP_016870205.1 | 5 | 953 | |||
| GenBank mRNA | AK127013.1 | - | 4 | 589 | ||

SNX30
Genomic Map