siRNA Details
| Gene Symbol: WDR38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045476.2 | NP_001038941.1 | 6 | 634 | ||
| NM_001276374.1 | NP_001263303.1 | 6 | 634 | |||
| NM_001276375.1 | NP_001263304.1 | 6 | 601 | |||
| NM_001276376.1 | NP_001263305.1 | 5 | 581 | |||
| XM_005251987.3 | XP_005252044.1 | 5 | 623 | |||

WDR38
Genomic Map