siRNA Details
| Gene Symbol: LCNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207510.3 | NP_997393.3 | 1 | 115 | ||
| GenBank mRNA | AK127160.1 | - | 1 | 115 | ||
| AK127236.1 | - | 1 | 1204 | |||
| BC137559.1 | - | 1 | 693 | 9021559 | ||
| BC137567.1 | - | 1 | 693 | 9021567 | ||

LCNL1
Genomic Map