siRNA Details
| Gene Symbol: MIR325HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110400.2 | 1 | 190 | |||
| NR_110401.2 | 1 | 190 | ||||
| NR_110402.2 | 1 | 190 | ||||
| NR_110403.2 | 1 | 190 | ||||
| NR_110404.2 | 1 | 190 | ||||
| NR_110405.2 | 1 | 190 | ||||
| NR_110406.2 | 1 | 190 | ||||
| GenBank mRNA | AK057746.1 | - | 1 | 309 | ||

MIR325HG
Genomic Map