siRNA Details
| Gene Symbol: LINC00890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033974.1 | 3 | 1299 | |||
| GenBank mRNA | BX537697.1 | - | 3 | 1288 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00890| Gene Symbol: LINC00890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033974.1 | 3 | 1299 | |||
| GenBank mRNA | BX537697.1 | - | 3 | 1288 | ||