siRNA Details
Gene Symbol: LINC01285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110393.1 | 2 | 244 | |||
GenBank mRNA | AK057519.1 | - | 2 | 244 | ||
DB165975.1 | - | 2 | 208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110393.1 | 2 | 244 | |||
GenBank mRNA | AK057519.1 | - | 2 | 244 | ||
DB165975.1 | - | 2 | 208 |