siRNA Details
| Gene Symbol: FAM224A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002161.1 | 2, 3 | 268 | |||
| GenBank mRNA | AI670915.1 | - | 1, 2 | 186 | ||
| BF476242.1 | - | 1, 2 | 192 | |||
| Gene Symbol: FAM224B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002160.1 | 2, 3 | 247 | |||
| GenBank mRNA | AI819212.1 | - | 1, 2 | 214 | ||

FAM224A
Genomic Map