siRNA Details
| Gene Symbol: FAM224A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002161.1 | 3 | 388 | |||
| GenBank mRNA | BF476242.1 | - | 1 | 72 | ||
| Gene Symbol: FAM224B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002160.1 | 3 | 367 | |||
| GenBank mRNA | AI819212.1 | - | 1 | 94 | ||

FAM224A
Genomic Map