siRNA Details
| Gene Symbol: LINC00643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015358.2 | 3 | 447 | |||
| GenBank mRNA | AK311468.1 | - | 3 | 651 | ||
| BX248751.1 | - | 3 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00643| Gene Symbol: LINC00643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015358.2 | 3 | 447 | |||
| GenBank mRNA | AK311468.1 | - | 3 | 651 | ||
| BX248751.1 | - | 3 | 325 | |||