siRNA Details
| Gene Symbol: ZNF738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027130.1 | 4 | 496 | |||
| GenBank mRNA | AK027579.1 | - | 4 | 438 | ||
| AK291002.1 | - | 4 | 435 | |||
| AK291193.1 | - | 4 | 403 | |||
| BC034499.1 | - | 4 | 504 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF738| Gene Symbol: ZNF738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027130.1 | 4 | 496 | |||
| GenBank mRNA | AK027579.1 | - | 4 | 438 | ||
| AK291002.1 | - | 4 | 435 | |||
| AK291193.1 | - | 4 | 403 | |||
| BC034499.1 | - | 4 | 504 | |||