siRNA Details
| Gene Symbol: SRRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013694.2 | NP_001013716.2 | 3 | 321 | ||
| XM_011530178.2 | XP_011528480.1 | 3 | 384 | |||
| XM_017028799.1 | XP_016884288.1 | 3 | 333 | |||
| GenBank mRNA | AK293916.1 | - | 4 | 301 | ||
| BC066962.1 | - | 4 | 304 | 4820477 | ||
| DA325985.1 | - | 3 | 316 | |||

SRRD
Genomic Map