siRNA Details
| Gene Symbol: VWC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080500.2 | NP_001073969.1 | 2 | 236 | ||
| GenBank mRNA | AB374231.1 | - | 1 | 106 | ||
| BC146903.1 | - | 2 | 236 | 9056831 | ||
| BC146911.1 | - | 2 | 236 | 9056839 | ||
| BC146931.1 | - | 2 | 236 | 9056859 | ||
| BC146935.1 | - | 2 | 236 | 9056863 | ||
| BC171841.1 | - | 2 | 236 | 9054495 | ||
| EF552207.1 | - | 1 | 109 | |||
| EU541473.1 | - | 1 | 109 | |||

VWC2L
Genomic Map