siRNA Details
| Gene Symbol: LOC105377752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010137.1 | XP_016865626.1 | 3 | 352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377752| Gene Symbol: LOC105377752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010137.1 | XP_016865626.1 | 3 | 352 | ||