siRNA Details
| Gene Symbol: LINC01000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024368.1 | 5 | 5850 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01000| Gene Symbol: LINC01000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024368.1 | 5 | 5850 | |||