siRNA Details
| Gene Symbol: LOC650226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029420.1 | 4 | 577 | |||
| GenBank mRNA | BF895050.1 | - | 2 | 189 | ||
| BF895055.1 | - | 3 | 493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC650226| Gene Symbol: LOC650226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029420.1 | 4 | 577 | |||
| GenBank mRNA | BF895050.1 | - | 2 | 189 | ||
| BF895055.1 | - | 3 | 493 | |||