siRNA Details
| Gene Symbol: LOC100507507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322263.1 | NP_001309192.1 | 1 | 701 | ||
| GenBank mRNA | AA029855.1 | - | 1 | 156 | ||
| AI394533.1 | - | 1 | 218 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507507| Gene Symbol: LOC100507507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322263.1 | NP_001309192.1 | 1 | 701 | ||
| GenBank mRNA | AA029855.1 | - | 1 | 156 | ||
| AI394533.1 | - | 1 | 218 | |||