siRNA Details
| Gene Symbol: LOC105375396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317905.1 | NP_001304834.1 | 2 | 450 | ||
| XM_017011679.1 | XP_016867168.1 | 2 | 172 | |||
| GenBank mRNA | KF595048.1 | - | 2 | 450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375396| Gene Symbol: LOC105375396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317905.1 | NP_001304834.1 | 2 | 450 | ||
| XM_017011679.1 | XP_016867168.1 | 2 | 172 | |||
| GenBank mRNA | KF595048.1 | - | 2 | 450 | ||