siRNA Details
| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY250185.1 | - | 1 | 169 | ||
| BC101193.2 | - | 1 | 169 | |||
| BC101194.2 | - | 1 | 169 | |||
| BC101195.1 | - | 1 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDFY3-AS2| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY250185.1 | - | 1 | 169 | ||
| BC101193.2 | - | 1 | 169 | |||
| BC101194.2 | - | 1 | 169 | |||
| BC101195.1 | - | 1 | 169 | |||