siRNA Details
| Gene Symbol: CTXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206833.3 | NP_996664.1 | 2 | 346 | ||
| GenBank mRNA | AK098834.1 | - | 2 | 346 | ||
| BC062702.1 | - | 2 | 328 | 4280819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTXN1| Gene Symbol: CTXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206833.3 | NP_996664.1 | 2 | 346 | ||
| GenBank mRNA | AK098834.1 | - | 2 | 346 | ||
| BC062702.1 | - | 2 | 328 | 4280819 | ||