siRNA Details
| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 1 | 575 | |||
| NR_027052.1 | 1 | 575 | ||||
| GenBank mRNA | AK096901.1 | - | 1 | 280 | ||
| BC110368.1 | - | 1 | 588 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP7-AS1| Gene Symbol: THAP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027051.1 | 1 | 575 | |||
| NR_027052.1 | 1 | 575 | ||||
| GenBank mRNA | AK096901.1 | - | 1 | 280 | ||
| BC110368.1 | - | 1 | 588 | |||