siRNA Details
| Gene Symbol: CELF2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046108.1 | 6 | 2088 | |||
| GenBank mRNA | BC046381.1 | - | 6 | 2090 | ||
| DB336852.1 | - | 2 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CELF2-AS2| Gene Symbol: CELF2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046108.1 | 6 | 2088 | |||
| GenBank mRNA | BC046381.1 | - | 6 | 2090 | ||
| DB336852.1 | - | 2 | 190 | |||