siRNA Details
| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143757.1 | NP_001137229.1 | 11 | 7182 | ||
| NM_030626.2 | NP_085129.1 | 11 | 7029 | |||
| GenBank mRNA | AL512722.1 | - | 1 | 1463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC27| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143757.1 | NP_001137229.1 | 11 | 7182 | ||
| NM_030626.2 | NP_085129.1 | 11 | 7029 | |||
| GenBank mRNA | AL512722.1 | - | 1 | 1463 | ||