siRNA Details
| Gene Symbol: TMEM9B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073431.1 | 2 | 541 | |||
| GenBank mRNA | BM724201.1 | - | 2 | 541 | ||
| DA631400.1 | - | 2 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM9B-AS1| Gene Symbol: TMEM9B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073431.1 | 2 | 541 | |||
| GenBank mRNA | BM724201.1 | - | 2 | 541 | ||
| DA631400.1 | - | 2 | 318 | |||