siRNA Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165030.2 | NP_001158502.1 | 2 | 494 | ||
| NM_015012.3 | NP_055827.1 | 2 | 494 | |||
| NR_028491.2 | 2 | 494 | ||||
| GenBank mRNA | AK303339.1 | - | 2 | 185 | ||
| BC035034.1 | - | 2 | 365 | 5259179 | ||
| BC044597.1 | - | 2 | 356 | 4811565 | ||
| BC064366.1 | - | 2 | 251 | |||
| BC091524.1 | - | 2 | 293 | 30512350 | ||
| BE673436.1 | - | 2 | 365 | |||
| BI596931.1 | - | 2 | 498 | |||
| BI829492.1 | - | 2 | 474 | |||
| BU752757.1 | - | 2 | 376 | |||
| D26067.1 | - | 2 | 300 | |||
| DA149513.1 | - | 2 | 360 | |||
| DR157551.1 | - | 2 | 347 | |||

TMEM41B
Genomic Map